Prohibition signs
Supplier: Brady
Certificates
About this item
Prohibition signs must have a black diagram encircled by a red band and dissected by a red diagonal bar.
- Laminated polyester
- Resistance to most chemical products and solvents
- Resistance against extreme temperatures
- Good weatherability and UV resistance
- Excellent adhesion to low surface energy surfaces
- Thickness: 0,1 mm
- Different diameters