Order Entry
Norway
ContactUsLinkComponent
Anti-Leu-Enkephalin Rabbit Polyclonal Antibody
Anti-Leu-Enkephalin Rabbit Polyclonal Antibody
Katalog # BSENR-1498-50
Supplier:  Biosensis
undefined
Anti-Leu-Enkephalin Rabbit Polyclonal Antibody
Katalog # BSENR-1498-50
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Leu-Enkephalin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Epitope:
    Human Leu-Enkephalin peptide (230 - 234)
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    Proenkephalin-A
  • Immunogen:
    Synthetic human Leu-Enkephalin peptide (230 - 234) conjugated to BSA.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Leu-Enkephalin is cleaved from the precursor Proenkephalin-A. Leu-Enkephalin is an endogenous opioid peptide that interacts with opioid receptors and produces analgesic effects.

Antibody recognizes Leu-Enkephalin in native applications. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user. Cross reactivity: Human; mouse; rat. Leu-Enkephalin is highly conserved so cross-reactivity with other species is expected. Cross-reactivity with other opioid peptides is as follows: with Met-enkephalin 0.93%; with beta-lipotropin 0.01%; with beta-endorphin 0.01%
Application Information:
A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Leu-Enkephalin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Leu-Enkephalin peptide (230 - 234)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat