Anti-Peptide YY Chicken Polyclonal Antibody
Katalog # BSENC-1536-100
Supplier: Biosensis
Specifications
- Antibody type:Primary
- Antigen name:Peptide YY
- Antigen symbol:PYY
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- Isotype:IgY
- Reactivity:Human,Rat,Mouse
- Epitope:C-terminus of Human mature Peptide YY (24-36 aa) .
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:PYY-I|Peptide tyrosine tyrosine|Peptide YY(3-36)|PYY
- Immunogen:A peptide from the C-terminus of human mature Peptide YY (24-36 aa) .
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Peptide YY (PYY) is secreted from endocrine cells in the lower small intestine, colon and pancreas. PYY inhibits exocrine pancreatic secretion, has a vasoconstrictory action and inhibitis jejunal and colonic mobility (Ref: SWISS-Prot).
Antibody recognizes Peptide YY in ELISAs, other applications have not been tested.
Application Information:
ELISA. Suggested dilution at 1:200 to 1:1,000 (coating antibody) and 1:2,000 to 1:5,000 (detection antibody). Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Peptide YY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminus of Human mature Peptide YY (24-36 aa) .
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat