Specifications
- Antibody type:Primary
- Antigen symbol:TGF-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- Isotype:IgY
- Reactivity:Pig,Human,Rat,Mouse
- Western blot:Yes
- Epitope:Human TGFbeta-1 amino acids 372-386
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:TGF-beta-1|TGFB|TGFB1
- Immunogen:A peptide from human and mouse Transforming growth factor beta-1 (372-386 aa).
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Transforming growth factor beta-1 (TGFB1) is a multi-functional cytokine with roles in proliferation, differentiation and other functions in many cell types. The secreted TGFB1 protein is cleaved into a latency-associated peptide (LAP) and a mature TGFB1 peptide (Ref: SWISS-Prot).
Antibody recognizes TGFB1 by western blot and ELISAs; works best with cell supernatants or fluids as TGFB1 is secreted. Other applications have not been tested.
Application Information:
WB and ELISA. Suggested dilution of 1:2,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: TGF-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human TGFbeta-1 amino acids 372-386
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat, Pig