Specifications
- Antibody type:Tag
- Antigen name:His tag
- Antigen symbol:HIS
- Clonality:Monoclonal
- Clone:BHISH8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Epitope:6-His
- Cross adsorption:No
- Format:Liquid
- Immunogen:Synthetic peptide containing 6 Histidine residues.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The 6X His tag is a short peptide sequence of 6 histidine residues. Epitopes such as the 6X His tag are often included with the target DNA at the time of cloning to produce fusion proteins containing the tag sequence. This allows anti-epitope tag antibodies such as this one to serve as a universal detection reagent for any recombinant protein containing this tag. Anti-epitope antibodies are a useful alternative to generating antibodies to identify a specific recombinant protein. The 6X His motif is often used as a tag on recombinant proteins to facilitate purification with immobilized metal-affinity chromatography.
His-tagged fusion proteins
Application Information:
Western Blotting (WB) and Immunoprecipitation (IP). Suggested starting dilutions are as follows: WB at dilutions of 1:1000, IP at dilutions of 1:200. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Tag
Antigen: His Tag
Clonality: Monoclonal
Clone: BHISH8
Conjugation: Unconjugated
Epitope: 6-His
Host: Mouse
Isotype:
Reactivity: