Specifications
- Antibody type:Primary
- Antigen name:Autophagy related 138
- Antigen symbol:ATG138
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:APG5
- Immunogen:A synthetic peptide corresponding to the C-terminal of human ATG-5L. No immunogenic carrier protein was conjugated to the immunogen. Instead, Adjukine B (see the Adjuvants in biosensis' product list) has been used to orchestrate/boost the immune response.
- Size:500 µg
- Pk:500 µG
Specifications
About this item
FUNCTION: Required for autophagy. Conjugates to ATG12 and associates with isolation membrane to form cup-shaped isolation membrane and autophagosome. The conjugate detaches from the membrane immediately before or after autophagosome formation is completed. FUNCTION: May play an important role in the apoptotic process, possibly within the modified cytoskeleton. Its expression is a relatively late event in the apoptotic process, occurring downstream of caspase activity. SUBCELLULAR LOCATION: Cytoplasm. Colocalizes with nonmuscle actin. ALTERNATIVE PRODUCTS: 2 named isoforms produced by alternative splicing. TISSUE SPECIFICITY: Ubiquitous. The mRNA is present at similar levels in viable and apoptotic cells, whereas the protein is dramatically highly expressed in apoptotic cells. INDUCTION: By apoptotic stimuli. PTM: Conjugated to ATG12; which is essential for autophagy, but is not required for association with isolation membrane. SIMILARITY: Belongs to the ATG5 family.
IHC and wb confirmed the specificity for ATG5.
Application Information:
IHC, immunofluorescence, WB (band at 49kDa) . A dilution of 1:200 to 1:1000 dilution is recommended for these applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: ATG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, not yet tested in other species