Bags, zip seal, black opaque, writing area, Minigrip®
Leverandør: Flexico
Sertifikat
Om dette produktet
PE bags, thickness 60 µm.
- Confidentiality of content due to opacity to light
- 3 white write-on panels
- Hanging hole
- Resealable, reusable, 100% recyclable
Made of polyethylene (LDPE). Zip seal designed for easy opening and high speed packing. The Minigrip® bag provides an excellent mechanical resistance and high temperature resistance (from –40 up to +60 °C). NOT suitable for food contact.