Order Entry
Norway
ContactUsLinkComponent
Anti-ANXA3 Rabbit Polyclonal Antibody
Anti-ANXA3 Rabbit Polyclonal Antibody
Catalog # ABNOH00000306-D01
Supplier:  Abnova
Anti-ANXA3 Rabbit Polyclonal Antibody
Catalog # ABNOH00000306-D01
Supplier:  Abnova

Specifications

  • Antistofftype:
    Primary
  • Antigen-navn:
    Annexin A3
  • Antigensymbol:
    ANXA3
  • Klonalitet:
    Polyclonal
  • Konjugering:
    Unconjugated
  • Vert:
    Rabbit
  • Immunutfelling:
    Yes
  • Reaktivitet:
    Human,
    Mouse
  • Western blot:
    Yes
  • Kyssabsorpsjon:
    No
  • Format:
    Liquid
  • Gen-ID:
    306
  • Antigen synonymer:
    ANXA3|Placental anticoagulant protein III|Annexin A3|ANX3|Lipocortin III|Annexin III|35-alpha calcimedin|PAP-III|Annexin-3|Inositol 1,2-cyclic phosphate 2-phosphohydrolase
  • Aminosyretall:
    1 to 323
  • Lagringsbuffer:
    No additive
  • Sekvens:
    MASIWVGHRGTVRDYPDFSPSVDAEAIQKAIRGIGTDEKMLISILTERSNAQRQLIVKEYQAAYGKELKDDLKGDLSGHFEHLMVALVTPPAVFDAKQLKKSMKGAGTNEDALIEILTTRTSRQMKDISQAYYTVYKKSLGDDISSETSGDFRKALLTLADGRRDESLKVDEHLAKQDAQILYKAGENRWGTDEDKFTEILCLRSFPQLKLTFDEYRNISQKDIVDSIKGELSGHFEDLLLAIVNCVRNTPAFLAERLHRALKGIGTDEFTLNRIMVSRSEIDLLDIRTEFKKHYGYSLYSAIKSDTSGDYEITLLKICGGDD
  • Lagringstemp:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    ANXA3 (NP_005130.1, 1 a.a. ~ 323 a.a) full-length human protein.
  • Str:
    100 μl
  • Fp:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against a full-length human ANXA3 protein.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ANXA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse