Order Entry
Norway
ContactUsLinkComponent
Anti-FSH Mouse Monoclonal Antibody [clone: 2F3]
Anti-FSH Mouse Monoclonal Antibody [clone: 2F3]
Catalog # ABNOMAB0029-M02
Supplier:  Abnova
Anti-FSH Mouse Monoclonal Antibody [clone: 2F3]
Catalog # ABNOMAB0029-M02
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Follicle Stimulating Hormone
  • Antigen symbol:
    FSH
  • Clonality:
    Monoclonal
  • Clone:
    2F3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Antigen synonyms:
    follicle-stimulating hormone beta subunit|follitropin|FSH-B|follitropin beta chain|follitropin subunit beta|FSH-beta|beta chain
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified Follicle Stimulating Hormone from urine of post-menopausal women.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against Follicle Stimulating Hormone.

Type: Primary
Antigen: FSH
Clonality: Monoclonal
Clone: 2F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human