Anti-SELL Mouse Monoclonal Antibody [clone: LT-TD180]
ABNOMAB0969
: Abnova
- Antibody type:Primary
- Antigen name:Selectin L
- Antigen symbol:SELL
- Clonality:Monoclonal
- Clone:LT-TD180
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:6402
- Antigen synonyms:LECAM1|CD62L|TQ1|LYAM1|LEU8|LNHR|LSEL|LAM1|PLNHR
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified SELL from peripheral blood leukocytes.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native SELL.
Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: LT-TD180
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human