Anti-Adenovirus E1A Mouse Monoclonal Antibody [clone: M73]
ABNOMAB1874
: Abnova
- Antibody type:Primary
- Antigen name:Adenovirus E1A
- Clonality:Monoclonal
- Clone:M73
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length adenovirus E1A.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against full length recombinant E1A.
Recommended Dilutions: Immunofluorescence: 2-5 ?g/ml; Immunoprecipitation: 1-2 ?g/ml
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: M73
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human