Anti-SOX9 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12736
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:SRY (sex determining region Y)-box 9
- Antigen symbol:SOX9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Chicken,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:6662
- Antigen synonyms:SRXX2|SRXY10|CMD1|SRA1|CMPD1
- Storage buffer:PBS, pH 7,2 (10% Proclin300)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal domain of human SOX9.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of SOX9.
Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; ELISA: 0.01-0.1 ?g/ml; Immunoprecipitation: 2-5 ?g/ml; Immunohistochemistry: 2-5 ?g/ml
Type: Primary
Antigen: SOX9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Human, Mouse, Rat