Anti-CDKN2B Mouse Monoclonal Antibody [clone: DCS114]
Catalog # ABNOMAB2030
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4)
- Antigen symbol:CDKN2B
- Clonality:Monoclonal
- Clone:DCS114
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1030
- Antigen synonyms:p15INK4b|TP15|P15|MTS2|CDK4I|INK4B
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human CDKN2B.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant CDKN2B.
Recommended Dilutions: Western Blot: 1 to 5 ?g /ml
Type: Primary
Antigen: CDKN2B
Clonality: Monoclonal
Clone: DCS114
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human