Anti-DYKDDDDK tag Monoclonal Antibody [clone: 29E4.G7.H11]
ABNOMAB2094
: Abnova
- Antibody type:Primary
- Antigen name:DYKDDDDK tag
- Antigen symbol:DYKDDDDK
- Clonality:Monoclonal
- Clone:29E4.G7.H11
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Storage buffer:20 mM potassium phosphate buffer, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to FLAG epitope tag.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against synthetic peptides of FLAG epitope tag.
Recommended Dilutions: ELISA: 1:160000; Western Blot: 1:2000-1:20000
Type: Primary
Antigen: DYKDDDDK tag
Clonality: Monoclonal
Clone: 29E4.G7.H11
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG
Reactivity: