Anti-RFP Polyclonal Antibody
ABNOPAB11376
: Abnova
- Antibody type:Primary
- Antigen name:Red Fluorescent Protein
- Antigen symbol:RFP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:DSRED
- Storage buffer:20 mM potassium phosphate buffer, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length mushroom polyp coral Discosoma RFP.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against full length recombinant RFP.
Recommended Dilutions: ELISA: 1:160000; Western Blot: 1:2000-1:10000; Immunohistochemistry: 1:2000-1:10000
Type: Primary
Antigen: RFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: