Anti-MAD1L1 Mouse Monoclonal Antibody [clone: P22.9B10.C12]
ABNOMAB8826
: Abnova
- Antistofftype:Primary
- Antigen-navn:MAD1 Mitotic Arrest Deficient-Like 1
- Antigensymbol:MAD1L1
- Klonalitet:Monoclonal
- Klon:P22.9B10.C12
- Konjugering:Unconjugated
- Vert:Mouse
- ImmunoFluorescens:Yes
- Immunutfelling:Yes
- Isotype:IgG2b
- Reaktivitet:Human
- Western blot:Yes
- Kyssabsorpsjon:No
- Format:Liquid
- Skjema:Ascites
- Gen-ID:8379
- Antigen synonymer:MAD1|TXBP181|PIG9|TP53I9
- Lagringsbuffer:PBS, pH 7,4 (0,02% sodium azide)
- Lagringstemp:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human MAD1L1.
- Str:100 μg
- Fp:100 µg
Mouse monoclonal antibody raised against recombinant MAD1L1.
Type: Primary
Antigen: MAD1L1
Clonality: Monoclonal
Clone: P22.9B10.C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human