Anti-VTN Mouse Monoclonal Antibody [clone: BE10]
ABNOMAB5220
: Abnova
- Antibody type:Primary
- Antigen name:Vitronectin
- Antigen symbol:VTN
- Clonality:Monoclonal
- Clone:BE10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:7448
- Antigen synonyms:Serum-spreading factor|Vitronectin V65 subunit|Somatomedin-B |Vitronectin|VTN|Vitronectin V10 subunit|S-protein|V75
- Storage buffer:PBS (1% BSA, 0,1% sodium azide)
- Storage temperature:Store at 4 °C.
- Immunogen:Native purified human VTN.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native VTN.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: VTN
Clonality: Monoclonal
Clone: BE10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human