Anti-H3K79me3 Polyclonal Antibody
Catalog # ABNOPAB14122
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:H3K79ME3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Format:Liquid
- Storage buffer:PBS (0,05% sodium azide, 0,05% procl300)
- Storage temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to region of histone H3 containing the trimethylated lysine 79 (H3K79me3).
- Tested applications:Dot Blot
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of H3K79me3.
Recommended Dilutions: ELISA: 1:500-1:1000; Dot Blot: 1:50000; Western Blot: 1:500 ChIP: 5 ?g/ChIP)
Type: Primary
Antigen: H3K79me3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human, Mouse