Anti-MT1A Rabbit Monoclonal Antibody [clone: N11-G]
ABNOMAB9787
: Abnova
- Antibody type:Primary
- Antigen name:Metallothionein 1A
- Antigen symbol:MT1A
- Clonality:Monoclonal
- Clone:N11-G
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:4489|4490|4493|4494|4495|4496|644314|4500|4499|450
- Storage buffer:20 mM Tris-HCl, pH 8.0 (10 mg/mL BSA, 0,05% sodium azide)
- Storage temperature:Store at –20 °C.
- Immunogen:A synthetic peptide corresponding to N-terminus of human MT1A, MT1B, MT1E, MT1F, MT1G, MT1H, MT1IP, MT1L, MT1M, MT2A.
- Size:50 μl
- Pk:50 µl
Rabbit monoclonal antibody raised against synthetic peptide of MT1A, MT1B, MT1E, MT1F, MT1G, MT1H, MT1IP, MT1L, MT1M, MT2A.
Recommended Dilutions: Western Blot: 1:1000; ELISA: 1:5000-1:10000
Type: Primary
Antigen: MT1A
Clonality: Monoclonal
Clone: N11-G
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse