Anti-AIRE Goat Polyclonal Antibody
ABNOPAB7187
: Abnova
- Antibody type:Primary
- Antigen name:Autoimmune Regulator
- Antigen symbol:AIRE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:326
- Antigen synonyms:autoimmune regulator|AIRE protein|AIRE|OTTHUMP00000109529|autoimmune polyendocrinopathy candidiasis ectodermal dystrophy|AIRE1|APECED|autoimmune regulator (automimmune polyendocrinopathy candidiasis ectodermal dystrophy)|PGA1|APSI|APS1|autoimmune regulator (APECED protein)|autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy)
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-KAKPPKKPESSAEQ
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human AIRE.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of AIRE.
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.3-1 ?g/ml
Type: Primary
Antigen: AIRE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse