Order Entry
Norway
ContactUsLinkComponent
 
Anti-BLM Rabbit Polyclonal Antibody
  ABNOPAB18272
 :  Abnova
Anti-BLM Rabbit Polyclonal Antibody
  ABNOPAB18272
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Bloom syndrome, RecQ helicase-like
  • Antigen symbol:
    BLM
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    641
  • Antigen synonyms:
    RECQL3|BS|RECQL2|RECQ2
  • Storage buffer:
    PBS, 150mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to human BLM.
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Rabbit polyclonal antibody raised against synthetic peptide of BLM.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Immunofluorescence: 1:500-1:1000; ELISA: 1:5000

Type: Primary
Antigen: BLM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human