Anti-CD2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RPA-2.10]
Supplier: Tonbo Biosciences
Certificates
About this item
The RPA-2.10 antibody reacts with human CD2, an approximately 50 kDa glycoprotein, and a member of the Ig superfamily. CD2, also known as LFA-2, is a receptor for CD58 in the human and is expressed on the cell surface of 80-90% of human peripheral blood lymphocytes, a subset of NK cells, and all mature T cells. CD2 mediates lymphocyte adhesion and is involved in T cell activation.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- 35-0029-T100
- 35-0029-T500
- 35-0029-T025
- Antigen symbol:
- CD2
- CD2
- CD2
- Antigen name:
- CD2 molecule
- CD2 molecule
- CD2 molecule
- Antigen synonyms:
- sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
- sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
- sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- RPA-2.10
- RPA-2.10
- RPA-2.10
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 914
- 914
- 914
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Yes
- Cross adsorption:
- No
- No
- No
- Storage temperature:
- Store undiluted at 2-8 °C, protect from light, do not freeze.
- Store undiluted at 2-8 °C, protect from light, do not freeze.
- Store undiluted at 2-8 °C, protect from light, do not freeze.