Order Entry
Norway
ContactUsLinkComponent
Anti-CD178 (Fas ligand) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: NOK-1]
Anti-CD178 (Fas ligand) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: NOK-1]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD178 (Fas ligand) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: NOK-1]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 50-9919-T100
  • 50-9919-T025
  • Antigen symbol:
  • FASLG
  • FASLG
  • Antigen name:
  • Fas Ligand
  • Fas Ligand
  • Antigen synonyms:
  • APT1LG1|APTL|TNFSF6|CD178|FASL|ALPS1B|CD95-L|CD95L
  • APT1LG1|APTL|TNFSF6|CD178|FASL|ALPS1B|CD95-L|CD95L
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • NOK-1
  • NOK-1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 356
  • 356
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • +2 °C to +8 °C, protected from light
  • +2 °C to +8 °C, protected from light

Specifications

Related Information

Frequently Bought Together