Anti-CD5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: UCHT2]
Supplier: Tonbo Biosciences
Certificates
About this item
The UCHT2 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein, that is expressed on most thymocytes, mature T cells, and a subset of B cells. CD5 is involved in modulating antigen receptor signaling in both T and B cells and binds to CD72, expressed on B cells.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Capuchin Monkey, Chimpanzee, Common Marmoset
Specifications
- Catalog No:
- 50-0059-T100
- 50-0059-T025
- Antigen symbol:
- CD5
- CD5
- Antigen name:
- CD5 molecule
- CD5 molecule
- Antigen synonyms:
- CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
- CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- UCHT2
- UCHT2
- Reactivity:
- Marmoset, Capuchin monkey, Chimpanzee, Human
- Marmoset, Capuchin monkey, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 921
- 921
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- +2 °C to +8 °C, protected from light
- +2 °C to +8 °C, protected from light