Anti-IL-10 Rat Monoclonal Antibody (In Vivo Ready™) [clone: JES5-2A5]
Supplier: Tonbo Biosciences
Certificates
About this item
The JES5-2A5 antibody is specific for mouse IL-10. IL-10 has numerous biological activities including the ability to inhibit both the synthesis of Th1 cytokines and proliferation of T cells. In the mouse, IL-10 is produced by phagocytic cells, conventional dendritic, cells, T cells, B cells, and NK cells. The JES5-2A5 antibody is a neutralizing antibody.
Type: Primary
Antigen: IL-10
Clonality: Monoclonal
Clone: JES5-2A5
Conjugation: In Vivo Ready™
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse
Specifications
- Catalog No:
- 40-7102-U100
- 40-7102-U500
- Antigen symbol:
- IL10
- IL10
- Antigen name:
- Interleukin 10
- Interleukin 10
- Antigen synonyms:
- IL10A|IL-10|GVHDS|TGIF|CSIF
- IL10A|IL-10|GVHDS|TGIF|CSIF
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- JES5-2A5
- JES5-2A5
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 16153
- 16153
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- +2 °C to +8 °C
- +2 °C to +8 °C