Order Entry
Norway
ContactUsLinkComponent
Anti-IFN gamma Rat Monoclonal Antibody (Biotin) [clone: XMG1.2]
Anti-IFN gamma Rat Monoclonal Antibody (Biotin) [clone: XMG1.2]
Supplier:  Tonbo Biosciences
Certificates
Anti-IFN gamma Rat Monoclonal Antibody (Biotin) [clone: XMG1.2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 30-7311-U025
  • 30-7311-U100
  • Antigen symbol:
  • IFNG
  • IFNG
  • Antigen name:
  • Interferon gamma
  • Interferon gamma
  • Antigen synonyms:
  • AF1|IFG|Interferon gamma transducerInterferon gamma|IFI|Interferon gamma receptor |IFN-gamma|Immune interferon
  • AF1|IFG|Interferon gamma transducerInterferon gamma|IFI|Interferon gamma receptor |IFN-gamma|Immune interferon
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • XMG1.2
  • XMG1.2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 15978
  • 15978
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ELISA:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • +2 °C...+8 °C, protected from light
  • +2 °C...+8 °C, protected from light

Specifications

Related Information