Anti-CD178 (Fas ligand) Mouse Antibody (APC (Allophycocyanine)) [clone: NOK-1]
Supplier: Tonbo Biosciences
Certificates
About this item
The NOK-1 antibody reacts with human CD178 (Fas ligand) in both membrane bound and soluble forms. Fas ligand is a 40 kDa transmembrane glycoprotein, a member of the TNF family, and is expressed by activated T and NK cells, neutrophils, and monocytes. Interactions between CD178 (Fas ligand) and CD95 (Fas) induce a program of apoptosis and play a key role in immune regulation and homeostasis. The extracellular domain of human CD178 can be cleaved from the surface by matrix metalloproteinases (MMPs) resulting in a 26 kDa soluble protein.
Type: Primary
Antigen: CD178 (Fas ligand)
Clonality:
Clone: NOK-1
Conjugation: APC (Allophycocyanine)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- 20-9919-T025
- 20-9919-T100
- Antigen symbol:
- FASLG
- FASLG
- Antigen name:
- Fas Ligand
- Fas Ligand
- Antigen synonyms:
- APT1LG1|APTL|TNFSF6|CD178|FASL|ALPS1B|CD95-L|CD95L
- APT1LG1|APTL|TNFSF6|CD178|FASL|ALPS1B|CD95-L|CD95L
- Antibody type:
- Primary
- Primary
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- NOK-1
- NOK-1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 356
- 356
- Isotype:
- IgG1
- IgG1
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Storage temperature:
- Store undiluted at 2-8 °C, protect from light, do not freeze.
- Store undiluted at 2-8 °C, protect from light, do not freeze.