Anti-VSIG8 Mouse Monoclonal Antibody [clone: 604CT14.1.2]
Catalog # ABGEAM2124A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen symbol:VSIG8
- Clonality:Monoclonal
- Clone:604CT14.1.2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Mouse
- Western blot:Yes
- Epitope:C-terminal
- Format:Ascites
- Gene ID:284677;391123
- Amino acid number:331 to 360
- Storage buffer:pH 7,4
- Molecular weight:43.891 kDa
- Sequence:GYPTQNVSRSLRRKYC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Shipping temperature:4 °C
- Immunogen:This VSIG8 antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 331-360 amino acids from the C-terminal region of human VSIG8.
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: VSIG8
Clonality: Monoclonal
Clone: 604CT14.1.2
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgM
Reactivity: Mouse