Anti-DMP1 Rabbit Polyclonal Antibody
Catalog # ABGEAP18965A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:dentin matrix acidic phosphoprotein 1
- Antigen symbol:DMP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Gene ID:1758
- Antigen synonyms:ARHP, ARHR, DMP-1
- Amino acid number:146 to 175
- Storage buffer:pH 7,4
- Molecular weight:55.782 kDa
- Sequence:TTSESRELDNEDRVDC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.57
- Shipping temperature:4 °C
- Immunogen:This DMP1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 146-175 amino acids from the N-terminal region of human DMP1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: DMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human