Anti-MAFK Mouse Monoclonal Antibody [clone: 1328CT786.105.125]
Catalog # ABGEAM8420B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog K
- Antigen symbol:MAFK
- Clonality:Monoclonal
- Clone:1328CT786.105.125
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:7975
- Antigen synonyms:P18|NFE2U
- Storage buffer:pH 7,4
- Molecular weight:17.523 kDa
- Sequence:MTTNPKPNKALKVKKEAGENAPVLSDDELVSMSVRELNQHLRGLTKEEVTRLKQRRRTLKNRGYAASCRIKRVTQKEELERQRVELQQEVEKLARENSSMRLELDALRSKYEALQTFARTVARGPVAPSKVATTSVITIVKSTELSSTSVPFSAAS
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5
- Shipping temperature:4 °C
- Immunogen:This MAFK antibody is generated from a mouse immunized with a KLH conjugated synthetic peptide between amino acids from the human region of human MAFK.
- Purification:Protein G Purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MAFK
Clonality: Monoclonal
Clone: 1328CT786.105.125
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human