Anti-CD144 Rabbit Polyclonal Antibody
Catalog # CAYM160840-500
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:CD144
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Bovine,Mouse
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:259 to 434
- Storage buffer:PBS, pH 7,2, containing 0,02% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human VE-cadherin amino acids 259-434
- Purification:purified by Protein A chromatography
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Type: Primary
Antigen: CD144
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse