Order Entry
Norway
ContactUsLinkComponent
Anti-TRAIL-R4 Rabbit Polyclonal Antibody
Anti-TRAIL-R4 Rabbit Polyclonal Antibody
Catalog # CAYM160755-1
Supplier:  Cayman Chemical
Anti-TRAIL-R4 Rabbit Polyclonal Antibody
Catalog # CAYM160755-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TRAIL-R4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Amino acid number:
    249 to 263
  • Storage buffer:
    PBS containing 0,02% sodium azide
  • Sequence:
    GGPERVHRVLFRRRS
  • Storage temperature:
    +4 °C
  • Shipping temperature:
    4 °C
  • Immunogen:
    human DcR2 precursor amino acids 249-263
  • Purification:
    purified by affinity chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: TRAIL-R4
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human