Order Entry
Norway
ContactUsLinkComponent
Anti-15-Hydroxy-PGDH Rabbit Polyclonal Antibody
Anti-15-Hydroxy-PGDH Rabbit Polyclonal Antibody
Catalog # CAYM160615-1
Supplier:  Cayman Chemical
Anti-15-Hydroxy-PGDH Rabbit Polyclonal Antibody
Catalog # CAYM160615-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    15-hydroxy PGDH
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Baboon,
    Human,
    Bovine,
    Guinea pig
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Amino acid number:
    92 to 105
  • Sequence:
    AGVNNEKNWEKTLQ
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    human NAD+-dependent 15-hydroxy PGDH amino acids 92-105
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: 15-hydroxy PGDH
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Baboon, Bovine, Guinea Pig, Human