Anti-Lp-PLA2 Rabbit Polyclonal Antibody
Catalog # CAYM160603-500
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:LP-PLA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Amino acid number:420 to 441
- Sequence:TNINTTNQHIMLQNSSGIEKYN
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human plasma PAF-AH C-terminal amino acids 420-441
- Purification:purified by affinity chromatography
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Type: Primary
Antigen: Lp-PLA2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human