Anti-GPR120 Rabbit Polyclonal Antibody
Catalog # CAYM14904-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:GPR120
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:Internal
- Cross adsorption:No
- Amino acid number:247 to 258
- Storage buffer:TBS, pH 7,4
- Sequence:SEITKASRKRLT
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human GPR120 amino acids 247-258
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: GPR120
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: Internal
Host: Rabbit
Isotype: IgG
Reactivity: Human