Anti-I kappaB-zeta Rabbit Polyclonal Antibody
Catalog # CAYM13926-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen name:I kappa B zeta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:684 to 699, 285 to 298
- Storage buffer:PBS, containing 0,2% gelatin and 0,05% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:synthetic peptide corresponding to mouse I kappaB-zeta amino acids 684-699 and 285-298
- Purification:purified by Protein G chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: I kappaB-zeta
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse