Anti-CASP3 Mouse Monoclonal Antibody [clone: 31A893]
Katalog # CAYM13911-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:CASP3
- Clonality:Monoclonal
- Clone:31A893
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Storage buffer:PBS containing 0,05% BSA and 0,05% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:recombinant full-length human caspase-3
- Purification:purified by Protein G chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: CASP3
Clonality: Monoclonal
Clone: 31A893
Conjugation: unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG
Reactivity: Human