Anti-HDAC-6 Rabbit Polyclonal Antibody
CAYM13499-1
- Antibody type:Primary
- Antigen symbol:HDAC-6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Amino acid number:1 to 16
- Storage buffer:PBS containing 0,05% BSA and 0,05% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:synthetic peptide corresponding to amino acids 1-16 of human HDAC6
- Purification:purified by Protein G chromatography
- Size:1 Ea
- Pk:1
Type: Primary
Antigen: HDAC-6
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse