Order Entry
Norway
ContactUsLinkComponent
Anti-AROS Rabbit Polyclonal Antibody
Anti-AROS Rabbit Polyclonal Antibody
Catalog # CAYM13496-1
Supplier:  Cayman Chemical
Anti-AROS Rabbit Polyclonal Antibody
Catalog # CAYM13496-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    AROS
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Amino acid number:
    1 to 50
  • Storage buffer:
    PBS containing 0,05% BSA and 0,05% sodium azide
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    synthetic peptide from human AROS within the region of amino acids 1-50
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: AROS
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human