Anti-DNMT Mouse Monoclonal Antibody [clone: 60B1220.1]
CAYM13479-1
- Antibody type:Primary
- Antigen symbol:DNMT
- Clonality:Monoclonal
- Clone:60B1220.1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Zebrafish,Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Amino acid number:637 to 650
- Storage buffer:PBS containing 0,05% BSA and 0,05% sodium azide
- Sequence:EKDDREDKENAFKR
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:synthetic peptide corresponding to amino acids 637-650 of human DNMT1
- Size:1 Ea
- Pk:1
Type: Primary
Antigen: DNMT
Clonality: Monoclonal
Clone: 60B1220.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Zebrafish