Anti-SUCNR1 Rabbit Polyclonal Antibody
Catalog # CAYM10928-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:Succinate receptor 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:New world monkey,Chimpanzee,Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:100 to 150
- Storage buffer:PBS containing 0,05% BSA and 0,05% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human SUCNR1 amino acids 100-150
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: SUCNR1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Human, New World Monkey, Rat