Anti-IL-33 Mouse Monoclonal Antibody [clone: IL333068A]
Supplier: Cayman Chemical
Certificates
About this item
Type: Primary
Antigen: IL-33
Clonality: Monoclonal
Clone: IL333068A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Specifications
- Catalog No:
- CAYM10810-50
- CAYM10810-100
- Antigen symbol:
- IL-33
- IL-33
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- IL333068A
- IL333068A
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- recombinant human IL-33
- recombinant human IL-33
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7,4
- PBS, pH 7,4
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- –20 °C
- –20 °C