Anti-CD25 PE Mouse Monoclonal Antibody [clone: 143-13]
Catalog # CAYM10339-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:CD25
- Clonality:Monoclonal
- Clone:143-13
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Storage buffer:TBS, pH 7,4, containing 50% glycerol, 5 mg/ml BSA, and 0,02% sodium azide
- Storage temperature:+4 °C
- Shipping temperature:4 °C
- Purification:purified
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: CD25 PE
Clonality: Monoclonal
Clone: 143-13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human