Anti-CD36 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: JC63.1]
Catalog # CAYM10009870-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:CD36
- Clonality:Monoclonal
- Clone:JC63.1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgA
- Reactivity:Human,Rat,Mouse
- Epitope:Full length
- Cross adsorption:No
- Storage buffer:PBS, pH 7,2, containing 50% glycerol, 0,5 mg/ml BSA, and 0,02% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:recombinant adenovirus expressing full length mouse CD36
- Purification:purified
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: JC63.1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Full length
Host: Mouse
Isotype: IgA
Reactivity: Human, Mouse, Rat