Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 649)
Catalog # ROCK610-443-002-05
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:DyLight® 649
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Western blot:Yes
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Secondary
Antigen: IgG(Mouse)-HL
Clonality: Polyclonal
Clone:
Conjugation: DyLight649
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse