Order Entry
Norway
ContactUsLinkComponent
Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 488)
Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # ROCK609-441-002-05
undefined
Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # ROCK609-441-002-05

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 488
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

Type: Secondary
Antigen: IgG(Human)-HL
Clonality: Polyclonal
Clone:
Conjugation: DyLight488
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human