Anti-IgG2a Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # ROCK610-103-041
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG2a
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Western blot:Yes
- ChIP:Yes
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Anti-Mouse IgG2a Peroxidase Secondary antibodies are suitable for ELISA, immunochemistry, western blotting as well as other anti IgG2a antibody based assays.
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!
Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse