Anti-GDF15 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # ROCK600-403-B07
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Growth differentiation factor 15
- Antigen symbol:GDF15
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Gene ID:9518
- Antigen synonyms:MIC-1|GDF-15|NAG-1|PTGFB|PDF|PLAB|MIC1
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:Anti-NAG-1 (C-terminal specific) antibody was prepared by repeated immunizations with a synthetic peptide corresponding to a region near the carboxy terminal end of human NAG-1 Protein. A residue of cysteine was added to facilitate coupling to KLH.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This affinity purified antibody is suitable for ELISA and western blotting of human NAG-1 Protein.
Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!
Type: Primary
Antigen: GDF15
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse