Anti-PCLO Rabbit Polyclonal Antibody
Catalog # ROCK600-401-G30
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:piccolo presynaptic cytomatrix protein
- Antigen symbol:PCLO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:27445
- Antigen synonyms:ACZ|PCH3
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,1% (w/v) Sodium Azide, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Piccolo Antibody was produced from whole rabbit serum prepared by repeated immunizations raised against full length Protein.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-Picollo Antibody is suitable for use in WB and IHC. Expect a band approximately ~550kDa corresponding to the molecular weight of Picollo. Multiple isoforms can be detected. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: PCLO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human