Anti-SOD2 Rabbit Polyclonal Antibody
ROCK600-401-G13
- Antibody type:Primary
- Antigen name:Superoxide Dismutase 2
- Antigen symbol:SOD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse,Hamster,Drosophila,Rabbit,Monkey,Xenopus,Sheep,Chicken,Human,Bovine,Guinea pig,Dog
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:24787
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,1% (w/v) Sodium Azide, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Mn SOD Antibody was produced from whole rabbit serum prepared by repeated immunizations raised against rat Mn SOD.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Anti-Mn SOD Antibody is suitable for use in WB, IP, ELISA, and IHC. Expect a band approximately ~25kDa Protein corresponding to the molecular mass of Mn superoxide dismutase (SOD) on SDS PAGE immunoblots.
Recommended Dilutions: ELISA: 1:200; Western Blot: 0.2µg/ml;
Type: Primary
Antigen: SOD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Bovine, Dog, Chicken, Dosophila, Guinea Pig, Hamster, Monkey, Rabbit, Sheep, Xenopus, Rat